Sodium dihydrogen phosphate dihydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:13472-35-0
:
Formula:NaH₂PO₄·2H₂O
MDL Number:MFCD00149209
:156.01 g/mol
Merck Index:12,08806
Boiling Pt:< 650 °C (1013 hPa)
Melting Pt:60 °C
Density:1.91 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-449-2
REACH:01-2119489796-13
Specification Test Results
- Assay:Min. 99.0 %
- pH (20°C; 5 %):4.2 - 4.5
- Heavy metals (as Pb):Max. 5 ppm
- Insolubility in water:Max. 50 ppm
- Total N (Nitrogen):Max. 20 ppm
- Cl (Chloride):Max. 10 ppm
- SO4 (Sulphate):Max. 100 ppm
- As (Arsenic):Max. 0.5 ppm
- Fe (Iron):Max. 5 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
28015_SODIUM DIHYDROGEN PHOSPHATE 2H2O ANALAR_BSE_REV0
BSE/TSE Certificates
spec_VWRC28015.261 spec_VWRC28015.261 spec_VWRC28015.294 spec_VWRC28015.294
Product specification
Frequently Bought Together