

Tetrasodium diphosphate decahydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:13472-36-1
:
Formula:Na₄P₂O₇·10H₂O
MDL Number:MFCD00149200
:446.06 g/mol
Melting Pt:990 °C
Density:1.82 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ300
EINECS:231-767-1
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:20 g/l (20 °C) (anhydrous substance)
- Melting Point:990 °C (anhydrous substance)
- Molar Mass:446.06 g/mol
- Bulk Density:650 kg/m3
- Density:1.82 g/cm3 (20 °C)
- pH:9.5 - 10.5 (50 g/l, H2O, 20 °C)
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.06591.0500_TDS_EN ProductPage_106591
Product specification
msds_106591
SDS
Frequently Bought Together