Order Entry
Switzerland
ContactUsLinkComponent
Sodium dichromate dihydrate ≥99%, Purum, Fluka™
Health
Corrosive
Toxic
Oxidizing
Environment
Sodium dichromate dihydrate ≥99%, Purum, Fluka™
Certificates
Health
Corrosive
Toxic
Oxidizing
Environment
Sodium dichromate dihydrate ≥99%, Purum, Fluka™
Certificates

Specification Test Results

  • Assay:
    Min. 99 %
  • Chloride (Cl):
    Max. 0.05 %
  • Sulfate (SO₄):
    Max. 0.5 %

Specification Test Results

Related Information

Frequently Bought Together