




Sodium dichromate dihydrate 99%
:7789-12-0
:
:Na₂Cr₂O₇·2H₂O
:MFCD00149166
:298.00 g/mol (261.97 anhy)
:12,08754
:357 °C
:2,35 g/cm³ (25 °C)
:Ambient
:234-190-3
:3087
:5.1,II
:GHS06
:GHS08
:GHS05
:GHS09
:GHS03
- Appearance:Orange Crystals or needles
- Titration Iodometric:≥98.5 %
- pH:3.5 to 3.9 (5 % solution)
- Chloride (Cl):≤100 ppm
- Sulfate (SO4):≤1000 ppm
- Insoluble matter:≤0.01 % (in water)
Frequently Bought Together