Order Entry
Switzerland
ContactUsLinkComponent
Sodium carbonate decahydrate, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
Irritant
Sodium carbonate decahydrate, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
Irritant
Sodium carbonate decahydrate, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    210 g/l (20 °C)
  • Melting Point:
    33 °C Elimination of water of crystallisation
  • Molar Mass:
    286.14 g/mol
  • Bulk Density:
    700 - 900 kg/m3
  • Density:
    1.44 g/cm3 (20 °C)
  • pH:
    11 - 12 (50 g/l, H2O, 25 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together