Lead(II) acetate trihydrate 99.5-103.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 26483.297
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay:99.5 - 103.0 %
- pH (20°C; 10 %):5.0 - 7.5
- Insolubility in water:Max. 50 ppm
- Cl (Chloride):Max. 5 ppm
- NO3 + NO2 (as NO3):Max. 100 ppm
- Ca (Calcium):Max. 50 ppm
- Cu (Copper):Max. 10 ppm
- Fe (Iron):Max. 10 ppm
- K (Potassium):Max. 50 ppm
- Na (Sodium):Max. 50 ppm
- Conforms to Reag. Ph.Eur.:Passes test
- Conforms to ACS:Passes test
Specification Test Results
About this item
:
:379.34 g/mol
Merck Index:13,05415
Boiling Pt:280 °C (1013 hPa)
Melting Pt:75 °C
Density:2.55 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-104-4
UN:1616
ADR:6.1,III
:GHS08
:GHS09
CAS Number:6080-56-4
Formula:(H₃CCOO)₂Pb·3H₂O
MDL Number:MFCD00150023