ContactUsLinkComponent
Silver nitrate 0.05 mol/l (0.05 N) in aqueous solution, AVS TITRINORM volumetric solution
Environment
Silver nitrate 0.05 mol/l (0.05 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier:  VWR Chemicals
Certificates
undefined
Environment
Silver nitrate 0.05 mol/l (0.05 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier:  VWR Chemicals
Certificates
Specification Test Results
  • Titer (20°C; real value 0.2 % accuracy):
    0.0499 - 0.0501 mol/l
  • NIST traceable:
    Confirmed