Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

(S)-(-)-Limonene, Supelco®

Flammable
Irritant
Environment
Supplier:   Merck
(S)-(-)-Limonene, Supelco®
Supplier:  Merck
Certificates
(S)-(-)-Limonene, Supelco®

2 Options Available Below

Product Details & Documents
Synonyms:  (-)-Limonene, (S)-1-Methyl-4-(prop-1-en-2-yl)cyclohex-1-ene, L(-)-Limonene, (S)-(-)-p-Mentha-1,8-diene, (S)-(-)-Dipentene, (S)-(-)-4-Isopropenyl-1-methyl-1-cyclohexene, (S)-(-)-1-Methyl-4-(1-methylvinyl)cyclohexene
  • CAS Number:5989-54-8
  • Formula:C₁₀H₁₆
  • MDL Number:MFCD00001558
  • Molecular Weight:136.24 g/mol
  • Merck Index:13,05515
  • Boiling Pt:175 °C (1013 hPa)
  • Melting Pt:–74 °C
  • Density:0.84 g/cm³ (20 °C)
  • Flash Pt:43 °C
  • Storage Temperature:Ambient
  • UN:2052
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ100
  • EINECS:227-815-6
Specifications for Product Family

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 mL
    Log in for availability
    Log in for availability
    1 * 1 mL
    1 * 1 mL - SFr. 66.90
    SFr. 66.90
    1 * 1 mL
       SFr. 66.90

    Specifications:
    More
    Specifications
    Cat. No.
    62128-1ML
  • Glass bottle
    5 mL
    Log in for availability
    Log in for availability
    1 * 5 mL
    1 * 5 mL - SFr. 200.00
    SFr. 200.00
    1 * 5 mL
       SFr. 200.00

    Specifications:
    More
    Specifications
    Cat. No.
    62128-5ML