Order Entry
Switzerland
ContactUsLinkComponent
 
Irritant
Calcium chloride 1.0 M CaCl2 ≥98% volumetric solution, Fluka™
Catalog # 21114-1L
CAS Number:  
Irritant
Calcium chloride 1.0 M CaCl2 ≥98% volumetric solution, Fluka™
Catalog # 21114-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    0.995 - 1.005 mol/l

Specification Test Results

About this item

 :
Boiling Pt:100 °C
Density:1.09 g/cm³
 :110.98 g/mol
EINECS:233-140-8
 :GHS07
Storage Temperature:Ambient
CAS Number:10043-52-4
Formula:CaCl₂
MDL Number:MFCD00010903