Order Entry
Switzerland
ContactUsLinkComponent
Supelco® Organic Reference Standard, (+)-Fenchone
Supelco® Organic Reference Standard, (+)-Fenchone
Catalog # 46208-5ML-F
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, (+)-Fenchone
Catalog # 46208-5ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • CAS No.:
    4695-62-9
  • Description:
    (+)-Fenchone
  • Seq:
    02
  • Storage conditions:
    10...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00070689
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (+)-1,3,3-Trimethyl-2-norbornanone; (1S)-1,3,3-Trimethylbicyclo[2.2.1]heptan-2-one
  • Melting point:
    5 - 7 °C (lit.)
  • Molecular weight:
    152,23
  • Density:
    0,945 g/ml at 20 °C (lit.)
  • Flash point:
    66 °C - Closed cup
  • Boiling point:
    63 - 65 °C/13 mmHg (lit.)

Specifications

About this item