ContactUsLinkComponent
Sulphuric acid 2 mol/l (4 N) in aqueous solution, AVS TITRINORM volumetric solution
Sulphuric acid 2 mol/l (4 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier:  VWR Chemicals
Certificates
undefined
Sulphuric acid 2 mol/l (4 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier:  VWR Chemicals
Certificates
Specification Test Results
  • Titer (20°C; real value 0.2 % accuracy):
    1.998 - 2.002 mol/l
  • NIST traceable:
    Confirmed