Order Entry
Switzerland
ContactUsLinkComponent
 
Sulphuric acid 0.5 mol/l (1 N) in aqueous solution
  ROTHX873.1
 :  Roth Carl
 :  
Sulphuric acid 0.5 mol/l (1 N) in aqueous solution
  ROTHX873.1
 :  Roth Carl
 :  

 

  • Pk:
    1
  • Seq:
    0001

 

 

 :
 :98.08 g/mol
 :1.01 g/cm³ (20 °C)
 :Ambient
 :231-639-5
 :2796
 :8,II
 :7664-93-9
 :H₂SO₄
 :MFCD00064589