ContactUsLinkComponent
Sulphuric acid standard solution 0.5 mol/l (1 N) in water
Corrosive
Sulphuric acid standard solution 0.5 mol/l (1 N) in water
Certificates
undefined
Corrosive
Sulphuric acid standard solution 0.5 mol/l (1 N) in water
Specification Test Results
  • Appearance:
    Clear colorless Solution
  • Strength:
    0.998 to 1.002 N (20°C)