ContactUsLinkComponent
Sodium hydroxide 1 mol/l (1 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
 :  Avantor
Sodium hydroxide 1 mol/l (1 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
 :  Avantor
  • Titer (mol/l):
    0.997 - 1.003
  • Chloride (Cl):
    max. 5
  • Iron (Fe):
    max. 0.5
  • Heavy Metals (as Pb):
    max. 1
Frequently Bought Together