Staining troughs, Schiefferdecker
BRND472500
: Brand
- Description:Staining troughs, Schiefferdecker
- W×D×H:70×85×45 mm
- Environmentally Preferable:
- Pk:10
Jars , made of soda-lime glass, with lid.
These staining trough hold up to 10 slides (76×26 mm), or 10 pairs of slides placed adjacently back-to-back.
Jars made of heavy-walled soda lime glass with a relatively high coefficient of expansion, and it should not exposed to rapid temperature changes, to avoid cracks caused by tensions.