Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Staining troughs, Hellendahl extended

Staining troughs, Hellendahl extended

Staining troughs acc. to Hellendahl, for 16 slides, extended, with lid, 55×60×105 mm
Catalog # BRND472700
Supplier:   Brand
Staining troughs, Hellendahl extended
Catalog # BRND472700
Supplier:  Brand
Staining troughs, Hellendahl extended
Staining troughs, Hellendahl extended

Staining troughs acc. to Hellendahl, for 16 slides, extended, with lid, 55×60×105 mm

Price
356.00 SFr.
Log in for availability
1 * 10 ST
Product Details & Documents

Soda-lime glass.

These staining trough hold up to 16 slides (76×26 mm), with widening towards the top.

Specifications
  • Description:
    Staining troughs acc. to Hellendahl, for 16 slides, extended, with lid
  • W×D×H:
    55×60×105 mm