Order Entry
Switzerland
ContactUsLinkComponent
Counting chambers, BLAUBRAND® Malassez
Counting chambers, BLAUBRAND® Malassez
Catalog # BRND719005
Supplier:  Brand
undefined
Counting chambers, BLAUBRAND® Malassez
Catalog # BRND719005
Supplier:  Brand

Specifications

  • Depth of chamber:
    0,2 mm
  • Type:
    Malassez
  • Description:
    Counting chamber, without clips, double ruling
  • Pk:
    1

Specifications

About this item

This counting chamber is commonly used for applications with nematodes.

  • Large rectangles of 0,25×0,20 = 0,05 mm²
  • Subdivision into 20 mini squares with an area of 0,0025 mm²
  • Rectangular ruling covering an surface of five mm²