ContactUsLinkComponent
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution
undefined
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution
  • Titer (20°C; real value 0.2 % accuracy):
    0.0998 to 0.1002 mol/l
  • Conforms to BDH 19145:
    Passes test
Frequently Bought Together