Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
Nitric acid 10 mol/l (10 N), Titripur® volumetric solution, Supelco®
  1.00630.1000
 :  Merck
 :  
 
 
Nitric acid 10 mol/l (10 N), Titripur® volumetric solution, Supelco®
  1.00630.1000
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Assay (HNO₃):
    48.0 - 50.0 %
  • Melting Point:
    -28 °C
  • Boiling Point:
    120 °C (1013 hPa)
  • Vapor Pressure:
    9.4 hPa (20 °C)
  • Density:
    1.38 g/cm3 (20 °C)
  • pH:
     - 1 (H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

 

 

 :
 :63.01 g/mol
 :120 °C (1013 hPa)
 :–28 °C
 :1.38 g/cm3 (20 °C)
 :Ambient
 :231-714-2
 :2031
 :8,II
 :M-Clarity™ quality level = MQ100
 :GHS05
 :GHS06
 :7697-37-2
 :HNO₃
 :MFCD00011349