Order Entry
Switzerland
ContactUsLinkComponent
Potassium dichromate 1/6 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Toxic
Health
Potassium dichromate 1/6 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 31396.291
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toxic
Health
Potassium dichromate 1/6 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 31396.291
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.1657 - 0.1663 mol/l
  • Appearance:
    Clear orange liquid
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
 :294.18 g/mol
Storage Temperature:Ambient
EINECS:231-906-6
UN:3287
ADR:6.1,III
 :GHS06
 :GHS08
CAS Number:7778-50-9
Formula:K₂Cr₂O₇
MDL Number:MFCD00011367