Order Entry
Switzerland
ContactUsLinkComponent
 
Periodic acid 0.1 mol/l (0.1 N) in aqueous solution
Catalog # 309746.1000
Supplier:  VWR Chemicals
CAS Number:  
undefined
Periodic acid 0.1 mol/l (0.1 N) in aqueous solution
Catalog # 309746.1000
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :227.94 g/mol
EINECS:233-937-0
CAS Number:10450-60-9
Formula:H₅IO₆
MDL Number:MFCD00011344