
Hydrochloric acid 1 mol/l (1 N), Titripur® Reag. Ph. Eur., Reag. USP volumetric solution, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
ADR:8,II
:M-Clarity™ quality level = MQ100
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS05
Storage Temperature:Ambient
Related Information
Frequently Bought Together