Order Entry
Switzerland
ContactUsLinkComponent
Hydrochloric acid concentrate for dilution to 0.1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Hydrochloric acid concentrate for dilution to 0.1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 32051.605
Supplier:  VWR Chemicals
CAS Number:  
undefined
Hydrochloric acid concentrate for dilution to 0.1 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 32051.605
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    60 mL
  • Seq:
    2385

Specifications

Specification Test Results

  • Titer (20°C):
    0.0995 - 0.1005 mol/l

Specification Test Results

About this item

 :
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8,III
 :Dose for preparation of 1 litre of solution.
 :36.46 g/mol
 :GHS05
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324