Order Entry
Switzerland
ContactUsLinkComponent
(±)-1,2-Propanediol ≥99%, TECHNICAL
(±)-1,2-Propanediol ≥99%, TECHNICAL
Supplier:  VWR Chemicals
Certificates
undefined
(±)-1,2-Propanediol ≥99%, TECHNICAL
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99 %
  • Refractive index (20°C):
    1.431 - 1.435

Specification Test Results

Frequently Bought Together