Orders LinkContactUsLinkComponent
Jars with cover, Nalgene®
Jars with cover, Nalgene®
   Sustainable Options Available
Jars with cover, Nalgene®
   Sustainable Options Available
Certificates
Jars with cover, Nalgene®
2 Options Available Below
Product Details & Documents
PP, translucent.
  • Straight sides and no neck making access and retrieval easy
  • Larger sizes make excellent waste crocks
  • Autoclavable
Ideal for storage and transport of solids.
Specifications for Product Family
Specifications
  • Catalog No:
  • NALG5352-0002
  • NALG5352-0004
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
Specifications
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Availability
    Price
  • 2200 ml
    132 mm
    193 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 128.00
    SFr. 128.00
    1 * 1 ST
       SFr. 128.00
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG5352-0002
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 4600 ml
    168 mm
    239 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 83.40
    SFr. 83.40
    1 * 1 ST
       SFr. 83.40
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG5352-0004
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact