ContactUsLinkComponent
Lid for 96-well PCR plates, Corning® Thermowell®
Lid for 96-well PCR plates, Corning® Thermowell®
Catalog # 734-1754
Supplier:  Corning
undefined
Lid for 96-well PCR plates, Corning® Thermowell®
Catalog # 734-1754
Supplier:  Corning
Specifications
  • Sterility:
    Non sterile
  • Colour:
    Clear
  • Description:
    Lid for 96-well PCR plate, PC
  • Pk:
    25
Specifications
About this item
Polycarbonate lids are thermally stable up to 135 °C and can be used on Corning® Thermowell™ 96 well polycarbonate plates (fits Cat. No. 6511) with oil overlays.
Lids are intended to be covers only and do not prevent evaporation.