Tubing, silicone, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Braided platinum-cured silicone tubing.
- Contains no phthalate plasticisers or peroxides to leach out into transported fluid
- Tasteless, odorless, non-cytotoxic, non-pyrogenic and non-hemolytic
- Autoclavable; also can be sterilized by gamma radiation, ETO and chemical procedures
- Durometer hardness: Shore A, 65
Tubing for a variety of pump and transfer applications, including pharmaceutical, laboratory, bioprocess manufacturing and food and beverage applications. This durable, high-purity tubing features an interwoven polyester braid for added safety, pressure and kink resistance.
Certifications: Meets FDA, USDA, U.S. Pharmacopeia Class VI, 24th Edition requirements for plastic materials; and 3A sanitary standards.
Specifications
- Catalog No:
- NALG8061-3130
- NALG8061-3070
- NALG8061-3190
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement