Order Entry
Switzerland
ContactUsLinkComponent
Orthophosphoric acid 25%, TECHNICAL
Corrosive
Orthophosphoric acid 25%, TECHNICAL
Catalog # 20629.463
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Orthophosphoric acid 25%, TECHNICAL
Catalog # 20629.463
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    25 L
  • Seq:
    5327

Specifications

Specification Test Results

  • Assay:
    24 - 26 %

Specification Test Results

About this item

 :
 :98 g/mol
Density:1.1462 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-633-2
UN:1805
ADR:8,III
 :GHS05
CAS Number:7664-38-2
Formula:H₃PO₄
MDL Number:MFCD00011340