Order Entry
Switzerland
ContactUsLinkComponent
Sodium hydroxide 50% in water for analysis
Corrosive
Sodium hydroxide 50% in water for analysis
Certificates
undefined
Corrosive
Sodium hydroxide 50% in water for analysis

Specification Test Results

  • Appearance:
    Clear colorless Viscous solution
  • Titration with HCl:
    49 to 51 %
  • Sodium carbonate (Na2CO3):
    ≤0.1 %
  • Sodium chloride (NaCl):
    ≤0.008 %
  • Iron (Fe):
    ≤0.0005 %

Specification Test Results

Related Information

Frequently Bought Together