Order Entry
Switzerland
ContactUsLinkComponent
Nalgene® Serum Vials, Crimp Finish
Nalgene® Serum Vials, Crimp Finish
Catalog # NALG322030-0010
   Low Manufacturing Impact
undefined
Nalgene® Serum Vials, Crimp Finish
Catalog # NALG322030-0010
   Low Manufacturing Impact

Specifications

  • Cap included:
    No
  • Cap Size:
    20 mm
  • Ø×H:
    24×50 mm
  • Packed:
    252/pack, 1260/case
  • Capacity:
    10,00 ml
  • Colour:
    Clear
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    1.260

Specifications

About this item

These vials are lighter in weight than glass, and are made for use with stoppers and caps. Protects against leakage, breakage, and contamination.

  • PETG
  • Noncytotoxic
  • Nonpyrogenic
  • Nonsterile
Certifications: ISO 13485:2003
Ordering information: Stoppers and caps not included.
Packaging: Packaged in trayless shrink-wrap modules.