ContactUsLinkComponent
Syringe filters optimised for scale up, Acrodisc®
Syringe filters optimised for scale up, Acrodisc®
Certificates
undefined
Syringe filters optimised for scale up, Acrodisc®
Certificates
Specifications
  • Catalog No:
  • 514-4238
  • 516-8971
  • 516-8973
  • 516-7717
  • Type:
  • Supor® EKV
  • Supor®
  • Fluorodyne® II
  • Posidyne®
  • Ø:
  • 25 mm
  • 25 mm
  • 25 mm
  • 25 mm
  • Max. operating pressure:
  • 5,4 bar at ambient temperature
  • 5,4 bar at ambient temperature
  • 5,4 bar at ambient temperature
  • 5,4 bar at ambient temperature
  • Max. temperature:
  • 60 °C at 4,1 bar
  • 60 °C at 4,1 bar
  • 60 °C at 4,1 bar
  • 60 °C at 4,1 bar
  • Water flow rate:
  • 130 ml/min, at 2,1 bar
  • 130 ml/min, at 2,1 bar
  • 130 ml/min (2,1 bar)
  • 77 ml/min (2,1 bar)