Erlenmeyer flasks, wide neck, with screw caps
Supplier: Brand
Certificates
About this item
PP, highly translucent.
- Very good chemical resistance
- Resistant to heat up to +121 °C (for 20 m), prolonged use +100 °C
- Also sealable with standard ground joint stoppers (stoppers not supplied)
With easy to read graduations. Autoclavable. The flasks feature a stopper bed and can also be sealed with standard ground joint polypropylene stoppers.
Frequently Bought Together