



Perchlorsäure 60%, EMSURE® ACS zur Analyse, Supelco®
Catalog # 1.00518.1001
Supplier: Merck
CAS Number:
Specifications
- Packung:Glasflasche
- VE:1 L
- Seq:5037
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Boiling Point:160 °C
- Density:1.53 g/cm3 (15 °C)
- pH:(H2O, 20 °C) strongly acid
Specification Test Results
About this item
:
:100.46 g/mol
Boiling Pt:160…181 °C (1013 hPa)
Melting Pt:–112.2 °C
Density:1.53…1.768 g/cm³ (20 °C)
Storage Temperature:Raumtemperatur
EINECS:231-512-4
UN:1873
ADR:5.1 (8),I
:M-Clarity™ quality level = MQ300
:GHS03
:GHS05
:GHS08
:GHS07
CAS Number:7601-90-3
Formula:HClO₄
MDL Number:MFCD00011325