

Hydrochloric acid 5 mol/l (5 N), AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Density:1.06 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8,III
:36.46 g/mol
:GHS07
:GHS05
Specification Test Results
- Titer (20°C; real value 0.2 % accuracy):4.99 - 5.01 mol/l
- NIST traceable:Confirmed
Specification Test Results
Frequently Bought Together