Hydrochloric acid 37% VLSI for microelectronic, J.T.Baker®
Catalog # 5367-03
Supplier: Avantor
CAS Number:
Specifications
- Pk:6 lb
- Seq:7059
- Environmentally Preferable:
Specifications
Specification Test Results
- For Microelectronic Use:37.0 - 38.0 %
- Store below 25°C:= 10
- Assay (as HCl) (by acid-base titrn):37.0 - 38.0 %
- Color (APHA):≤ 10
- Residue after Ignition:≤ 3 ppm
- Extractable Organic Substances:≤ 3 ppm
- Bromide (Br):≤ 0.005 %
- Free Halogen (as Cl₂):Passes Test
- Ammonium (NH₄):≤ 1 ppm
- Phosphate (PO₄):≤ 0.05 ppm
- Sulfate (SO₄):≤ 0.3 ppm
- Sulfite (SO₃):≤ 0.8 ppm
- Trace Impurities - Aluminum (Al):≤ 100.0 ppb
- Arsenic and Antimony (as As):≤ 5.0 ppb
- Trace Impurities - Barium (Ba):≤ 20.0 ppb
- Trace Impurities - Beryllium (Be):≤ 10.0 ppb
- Trace Impurities - Bismuth (Bi):≤ 20.0 ppb
- Trace Impurities - Boron (B):≤ 20.0 ppb
- Trace Impurities - Cadmium (Cd):≤ 5.0 ppb
- Trace Impurities - Calcium (Ca):≤ 100.0 ppb
- Trace Impurities - Chromium (Cr):≤ 50 ppb
- Trace Impurities - Cobalt (Co):≤ 5.0 ppb
- Trace Impurities - Copper (Cu):≤ 5.0 ppb
- Trace Impurities - Gallium (Ga):≤ 20.0 ppb
- Trace Impurities - Germanium (Ge):≤ 20 ppb
- Trace Impurities - Gold (Au):≤ 20 ppb
- Heavy Metals (as Pb):≤ 100 ppb
- Trace Impurities - Iron (Fe):≤ 50.0 ppb
- Trace Impurities - Lead (Pb):≤ 25.0 ppb
- Trace Impurities - Lithium (Li):≤ 50.0 ppb
- Trace Impurities - Magnesium (Mg):≤ 50.0 ppb
- Trace Impurities - Manganese (Mn):≤ 5.0 ppb
- Trace Impurities - Molybdenum (Mo):≤ 10.0 ppb
- Trace Impurities - Nickel (Ni):≤ 10.0 ppb
- Trace Impurities - Niobium (Nb):≤ 10.0 ppb
- Trace Impurities - Potassium (K):≤ 100 ppb
- Trace Impurities - Silicon (Si):≤ 100.0 ppb
- Trace Impurities - Silver (Ag):≤ 20.0 ppb
- Trace Impurities - Sodium (Na):≤ 100.0 ppb
- Trace Impurities - Strontium (Sr):≤ 20.0 ppb
- Trace Impurities - Tantalum (Ta):≤ 10.0 ppb
- Trace Impurities - Thallium (Tl):≤ 20.0 ppb
- Trace Impurities - Tin (Sn):≤ 50 ppb
- Trace Impurities - Titanium (Ti):≤ 10.0 ppb
- Trace Impurities - Vanadium (V):≤ 10.0 ppb
- Trace Impurities - Zinc (Zn):≤ 50 ppb
- Trace Impurities - Zirconium (Zr):≤ 10.0 ppb
- Particle Count - 0.5 µm and greater:≤ 50 par/ml
- Particle Count - 1.0 µm and greater:≤ 10 par/ml
Specification Test Results
About this item
Synonyms: Hydrochloric acid solution, Muriatic acid
:
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–30 °C
Density:1.18 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1789
ADR:8,II
:Management of Change (MOC) category = R
:36.46 g/mol
EINECS:231-595-7
:GHS05
:GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324