Vented caps for Falcon® flasks
Anbieter: Corning
Zertifikate
Über diesen Artikel
PE with integral hydophobic 0,2 µm microporous membrane filter vent.
Vented caps minimise contamination associated with standard open incubation. Gases required for cell growth and metabolism pass freely through the vent, while microorganisms do not.
Technische Daten
- Catalog No:
- 734-1068
- 734-1067
- 734-1066
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 - Bedford
- ISO 14001 - Bedford
- ISO 14001 - Bedford
Technische Daten
Frequently Bought Together