Order Entry
Switzerland
ContactUsLinkComponent
Formaldehyde 37% (w/w) in water stabilised
Toxic
Health
Corrosive
Flammable
Formaldehyde 37% (w/w) in water stabilised
Certificates
undefined
Toxic
Health
Corrosive
Flammable
Formaldehyde 37% (w/w) in water stabilised

Specification Test Results

  • Appearance:
    Clear solution
  • Color scale:
    ≤10 APHA
  • Stabilizer:
    5 to 15 % methanol
  • Titration:
    35.0 to 39.0 %
  • Free acid (HCOOH):
    ≤0.1 %

Specification Test Results

Related Information

Frequently Bought Together