


Benzene-[D₆] (99.96% D) for NMR spectroscopy, Sigma-Aldrich®
Catalog # 1.01766.0009
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass ampoule
- Pk:7,5 mL
- Seq:2260
Specifications
About this item
:
:84.07 g/mol
Boiling Pt:79 °C (1013 hPa)
Melting Pt:6.7 °C
Density:0.95 g/cm³ (20 °C)
Flash Pt:–11 °C (closed cup)
Storage Temperature:Ambient
EINECS:214-061-8
UN:1114
ADR:3,II
:M-Clarity™ quality level = MQ200
:GHS02
:GHS08
:GHS07
CAS Number:1076-43-3
Formula:C₆D₆
MDL Number:MFCD00003010