Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
n-Heptane-[D16]
n-Heptane-[D₁₆]
n-Heptane-[D₁₆]
n-Heptane-[D₁₆]
8.15039.0005
:
Merck
:
n-Heptane-[D₁₆]
8.15039.0005
:
Merck
:
Pack type:
Glass bottle
Pk:
5 mL
Seq:
2210
:
:
116.08 g/mol
:
1206
:
3,II
:
GHS02
:
GHS08
:
GHS07
:
GHS09
:
Ambient
:
33838-52-7
:
D₃C(CD₂)₅CD₃
:
MFCD00062744