


Methanol-[D₄] (99.8% D), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:811-98-3
:
Formula:D₃COD
MDL Number:MFCD00044637
:36.01 g/mol
Boiling Pt:65.3 °C (1013 hPa)
Melting Pt:–99 °C
Density:0.888 g/cm³ (20 °C)
Flash Pt:11 °C
Storage Temperature:Ambient
UN:1230
ADR:3 (6.1),II
:M-Clarity™ quality level = MQ200
EINECS:212-378-6
:GHS02
:GHS06
:GHS08
Frequently Bought Together