

Chloroform-[D₁] (99.8% D) stabilised, MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:865-49-6
:
Formula:CDCl₃
MDL Number:MFCD00000827
:120.38 g/mol
Boiling Pt:60.8 °C (1013 hPa)
Melting Pt:–64.1 °C
Density:1.5 g/cm³ (20 °C)
EINECS:212-742-4
UN:1888
ADR:6.1,III
:M-Clarity™ quality level = MQ200
:Stabilised with silver
:GHS06
:GHS08
Storage Temperature:Refrigerator
Frequently Bought Together