

Chloroform-[D₁], dehydrated (max. 0.01% H₂O) (99.8% D) for NMR spectroscopy
FLUOD-307-H
:
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
:
:120.38 g/mol
:60.8 °C (1013 hPa)
:–64.1 °C
:1.5 g/cm³ (20 °C)
:Refrigerator
:212-742-4
:1888
:6.1,III
:GHS08
:GHS07
:865-49-6
:CDCl₃
:MFCD00000827