

1,1,2,2-Tetrachloroethane-[D₂] (99.5% D), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:33685-54-0
:
Formula:Cl₂CDCDCl₂
MDL Number:MFCD00037672
:169.83 g/mol
Boiling Pt:146 °C (1013 hPa)
Melting Pt:–43 °C
Density:1.6118 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:251-634-1
ADR:6.1,II
:M-Clarity™ quality level = MQ200
UN:1702
:GHS06
:GHS09
Frequently Bought Together