

Acetone-[D₆] (99.9% D), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
1.00021.0100
: Merck
:
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
:
:64.03 g/mol
:55.5 °C (1013 hPa)
:–95.4 °C
:0.79 g/cm³ (20 °C)
:–19 °C (closed cup)
:Ambient
:211-563-9
:1090
:3,II
:M-Clarity™ quality level = MQ200
:GHS02
:GHS07
:666-52-4
:D₃CC(O)CD₃
:MFCD00044635