Xylene (mixture of isomers), BAKER, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:1330-20-7
:
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00077264
:106.17 g/mol
Merck Index:13,10136
Boiling Pt:140 °C (1013 hPa)
Melting Pt:–34 °C
Density:0.86…0.88 g/cm³ (20 °C)
Flash Pt:25 °C
Storage Temperature:Ambient
EINECS:215-535-7
UN:1307
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:passes test
- Assay:min. 98 %
- Color:max. 10
Specification Test Results
Frequently Bought Together