Xylene (mixture of isomers), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:1330-20-7
:
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00077264
:106.17 g/mol
Merck Index:13,10136
Boiling Pt:140 °C (1013 hPa)
Melting Pt:–34 °C
Density:0.86…0.88 g/cm³ (20 °C)
Flash Pt:25 °C
Storage Temperature:Ambient
EINECS:215-535-7
UN:1307
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Assay:min. 98.5 %
- Color (APHA):max. 10
- Ethylbenzene:max. 25 %
- Residue after Evaporation:max. 0.002 %
- Sulfur Compounds (as S):max. 0.003 %
- Substances Darkened by H2SO4:passes test
- Water (H2O):max. 0.03 %
- Aluminium (Al):max. 0.5
- Boron (B):max. 0.02
- Barium (Ba):max. 0.1
- Calcium (Ca):max. 0.5
- Cadmium (Cd):max. 0.05
- Cobalt (Co):max. 0.02
- Chromium (Cr):max. 0.02
- Copper (Cu):max. 0.02
- Iron (Fe):max. 0.1
- Magnesium (Mg):max. 0.1
- Manganese (Mn):max. 0.02
- Nickel (Ni):max. 0.02
- Lead (Pb):max. 0.1
- Tin (Sn):max. 0.1
- Zinc (Zn):max. 0.1
Specification Test Results
Frequently Bought Together