Order Entry
Switzerland
ContactUsLinkComponent
Xylene (mixture of isomers), J.T.Baker®
Xylene (mixture of isomers), J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Xylene (mixture of isomers), J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Benzene:
    max. 0.02 %
  • Color (APHA):
    max. 20
  • Density (g/ml) at 20°C:
    0.862 - 0.872

Specification Test Results

Related Information

Frequently Bought Together