Xylene (mixture of isomers) ≥98.5% (isomers plus ethylbenzene) ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1330-20-7
:
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00077264
:106.17 g/mol
Merck Index:14,10081
Boiling Pt:137…144 °C
Melting Pt:< –25 °C
Density:0.860
Flash Pt:29 °C (84 °F)
Storage Temperature:Ambient
EINECS:215-535-7
UN:1307
ADR:3,III
:GHS02
:GHS07
Related Information
Frequently Bought Together