Xylene (mixture of isomers) 99+%, Electronic Grade for residue analysis
Catalog # ACRO327950010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.02 %
- GC:≥99.0 % Xylene isomers + Ethylbenzene
- Color scale:≤10 APHA
- Residue after evaporation:≤0.0005 %
- Chloride (Cl):≤2 ppm
- Arsenic (As):≤10 ppb
- Barium (Ba):≤20 ppb
- Copper (Cu):≤10 ppb
- Iron (Fe):≤50 ppb
- Heavy metals (as Pb):≤1 ppm
- Acidity (HCl):≤0.001 %
Specification Test Results
About this item
:
:106.17 g/mol
Merck Index:13,10136
Boiling Pt:140 °C (1013 hPa)
Melting Pt:–34 °C
Density:0.865 g/cm³
Flash Pt:23 °C
EINECS:215-535-7
UN:1307
ADR:3,III
:GHS02
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:1330-20-7
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00077264