


Xylene (mixture of isomers) 96% (isomers plus ethylbenzene), pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1330-20-7
:
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00077264
:106.17 g/mol
Merck Index:13,10136
Boiling Pt:140 °C (1013 hPa)
Melting Pt:–34 °C
Density:0.865 g/cm³
Flash Pt:23 °C
Storage Temperature:Ambient
EINECS:215-535-7
UN:1307
ADR:3,III
:GHS02
:GHS07
:GHS08
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4940 to 1.5000 (20°C, 589 nm)
- GC:≥95.0 % Xylene isomers + Ethylbenzene
- Color scale:≤20 APHA
Specification Test Results
Frequently Bought Together